missing translation for 'onlineSavingsMsg'
Learn More

(S)-Metolachor, MedChemExpress

Product Code. 16493266
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
50 mg
Unit Size:
25mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493266 25 mg 25mg
16441127 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16493266 Supplier Medchem Express Supplier No. HY117279
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress (S)-Metolachor, a derivative of aniline, is a major pesticide in use.

(S)-Metolachor, a derivative of aniline, is a major pesticide in use.

Specifications

Chemical Name or Material (S)-Metolachor
CAS 87392-12-9
Color Colorless
Purity Grade Notes Research
Molecular Formula C15H22ClNO2
Quantity 25 mg
Solubility Information DMSO : 100 mg/mL (352.37 mM; Need ultrasonic)
SMILES O=C(N(C1=C(C)C=CC=C1CC)[C@@H](C)COC)CCl
Molecular Weight (g/mol) 283.79
Formula Weight 283.79
Percent Purity 98.5%
Grade Research
Physical Form Liquid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.