missing translation for 'onlineSavingsMsg'
Learn More

S-Methylisothiouronium sulfate, 98+%

Product Code. 11428883
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
2500 g
Unit Size:
100g
2500g
250g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428883 100 g 100g
11453980 250 g 250g
11438883 500 g 500g
11448883 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11428883 Supplier Thermo Scientific Alfa Aesar Supplier No. A11044.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 867-44-7 | C4H14N4O4S3 | 278.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 867-44-7
Molecular Formula C4H14N4O4S3
Molecular Weight (g/mol) 278.36
MDL Number MFCD00013055
InChI Key BZZXQZOBAUXLHZ-UHFFFAOYSA-N
Synonym s-methylisothiourea hemisulfate, 2-methyl-2-thiopseudourea hemisulfate, carbamimidothioic acid, methyl ester, sulfate 2:1, s-methylisothiourea sulfate 2:1, s-methylthiouronium sulfate 2:1, bis 2-methylisothiouronium sulphate, methyl carbamimidothioate sulfate 2:1, methylcarbamimidothioate sulfate 2:1, s-methylisothiourea, sulfate, 2-methyl-2-thiopseudourea sulfate 2:1
PubChem CID 13347
IUPAC Name methyl carbamimidothioate;sulfuric acid
SMILES CSC(=N)N.CSC(=N)N.OS(=O)(=O)O

Specifications

Melting Point ∼235°C (decomposition)
Density 1.28
Quantity 100 g
Beilstein 3917217
Sensitivity Hygroscopic
Formula Weight 278.38
Percent Purity ≥98%
Chemical Name or Material S-Methylisothiouronium sulfate

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.