missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-alpha-Methylbenzyl isocyanate, 98%

Product Code. 10236260 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
Packaging:
Glass bottle
Unit Size:
1mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10236260 5 mL Glass bottle 5mL
10657934 1 mL Glass bottle 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10236260 Supplier Thermo Scientific Acros Supplier No. 205310050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14649-03-7 | C9H9NO | 147.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chiral derivatization reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14649-03-7
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00002036
InChI Key JJSCUXAFAJEQGB-QMMMGPOBSA-N
Synonym s---1-phenylethyl isocyanate, 1s-1-isocyanatoethyl benzene, s---alpha-methylbenzyl isocyanate, benzene, 1s-1-isocyanatoethyl, isocyanic acid s---a-methylbenzyl ester, s-alpha-methylbenzyl isocyanate, s---1-phenylethylisocyanate, isocyanic acid s---alpha-methylbenzyl ester, isocyanic acid s---methylbenzyl ester, 1-isocyanatoethyl benzene #
PubChem CID 2724173
IUPAC Name [(1S)-1-isocyanatoethyl]benzene
SMILES CC(C1=CC=CC=C1)N=C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0500g/mL
Boiling Point 55.0°C to 56.0°C (2.5 mmHg)
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH(CH3)NCO
Refractive Index 1.5130 to 1.5150
Quantity 5 mL
Specific Gravity 1.05
Specific Rotation Condition −10° (20°C neat)
Specific Rotation − 10.00
Solubility Information Solubility in water: decomposes
Formula Weight 147.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (S)-(-)-α-Methylbenzyl isocyanate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • (S)-(-)-a-Methylbenzyl isocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.