missing translation for 'onlineSavingsMsg'
Learn More

S-Methyl thioacetate, 98+%

Product Code. 11489563
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489563 100 g 100g
11469563 5 g 5g
11479563 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489563 Supplier Thermo Scientific Alfa Aesar Supplier No. A12400.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1534-08-3 | C3H6OS | 90.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1534-08-3
Molecular Formula C3H6OS
Molecular Weight (g/mol) 90.14
MDL Number MFCD00014989
InChI Key OATSQCXMYKYFQO-UHFFFAOYSA-N
Synonym s-methyl thioacetate, methylthioacetate, ethanethioic acid, s-methyl ester, methanethiol acetate, thioacetic acid s-methyl ester, methyl thiolacetate, methyl ethanethioate, unii-pf2d4mwx79, ch3c o sch3, acetic acid, thio-, s-methyl ester
PubChem CID 73750
ChEBI CHEBI:51280
IUPAC Name S-methyl ethanethioate
SMILES CC(=O)SC

Specifications

Density 1.024
Boiling Point 97°C to 99°C
Flash Point 12°C (54°F)
Odor Unpleasant
Quantity 100 g
UN Number UN1993
Formula Weight 91.14
Percent Purity ≥98%
Chemical Name or Material S-Methyl thioacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.