missing translation for 'onlineSavingsMsg'
Learn More

S-Methyl-N,N-diethylthiocarbamate Sulfone, TRC

Product Code. 17981275
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17981275 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17981275 Supplier Toronto Research Chemicals Supplier No. TRCM3017505MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 155514-79-7
Molecular Formula C6 H13 N O3 S
Quantity 5 mg
InChI Formula InChI=1S/C6H13NO3S/c1-4-7(5-2)6(8)11(3,9)10/h4-5H2,1-3H3
SMILES CCN(CC)C(=O)S(=O)(=O)C
IUPAC Name N,N-diethyl-1-methylsulfonylformamide
Molecular Weight (g/mol) 179.24
Formula Weight 179.0616
Chemical Name or Material S-Methyl-N,N-diethylthiocarbamate Sulfone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.