missing translation for 'onlineSavingsMsg'
Learn More

S-Methyl-L-cysteine, MedChemExpress

Product Code. 16478866
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16478866 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16478866 Supplier Medchem Express Supplier No. HYB2188

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress S-Methyl-L-cysteine is a natural product that acts as a substrate in the catalytic antioxidant system mediated by methionine sulfoxide reductase A (MSRA), with antioxidative, neuroprotective, and anti-obesity activities.

S-Methyl-L-cysteine is a natural product that acts as a substrate in the catalytic antioxidant system mediated by methionine sulfoxide reductase A (MSRA), with antioxidative, neuroprotective, and anti-obesity activities.

Specifications

Chemical Name or Material S-Methyl-L-cysteine
CAS 1187-84-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C4H9NO2S
Synonym L-S-Methylcysteine
Solubility Information H2O : 30 mg/mL (221.93 mM; Need ultrasonic and warming)
SMILES N[C@@H](CSC)C(O)=O
Molecular Weight (g/mol) 135.18
Quantity 100 mg
Formula Weight 135.18
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.