missing translation for 'onlineSavingsMsg'
Learn More

(S)-MCPG, MedChemExpress

Product Code. 16462576
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16462576

Brand: Medchem Express HY100406

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (S)-MCPG ((+)-MCPG) is a potent group I/II metabotropic glutamate receptor (mGluRs) antagonist and the active isomer of (RS)-MCPG (HY-100371). (S)-MCPG can be used for the study of the function of mGluRs in spatial learning.

(S)-MCPG ((+)-MCPG) is a potent group I/II metabotropic glutamate receptor (mGluRs) antagonist and the active isomer of (RS)-MCPG (HY-100371). (S)-MCPG can be used for the study of the function of mGluRs in spatial learning.

Specifications

Chemical Name or Material (S)-MCPG
Physical Form Solid
CAS 150145-89-4
Color Off-White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C10H11NO4
For Use With (Application) Neuroscience-Neuromodulation
Synonym (+)-MCPG
Solubility Information DMSO : 1 mg/mL (4.78 mM; Need ultrasonic)
SMILES OC(C1=CC=C([C@](C)(N)C(O)=O)C=C1)=O
Molecular Weight (g/mol) 209.2
Formula Weight 209.2
Percent Purity 99.61%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.