missing translation for 'onlineSavingsMsg'
Learn More

S-(+)-Marmesin, MedChemExpress

Product Code. 16473946
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16473946 25 mg 25mg
16459268 10 mg 10mg
16441688 5 mg 5mg
16434937 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16473946 Supplier Medchem Express Supplier No. HYN2176

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress S-(+)-Marmesin is a natural coumarin, exhibiting COX-2/5-LOX dual inhibitory activity.

S-(+)-Marmesin is a natural coumarin, exhibiting COX-2/5-LOX dual inhibitory activity.

Specifications

Chemical Name or Material S-(+)-Marmesin
CAS 13849-08-6
Color White
Purity Grade Notes Research
Molecular Formula C14H14O4
Quantity 25 mg
Synonym (+)-Marmesin (S)-Marmesin
Solubility Information DMSO : 2 mg/mL (8.12 mM; Need ultrasonic)
SMILES O=C1C=CC2=CC3=C(O[C@H](C(C)(O)C)C3)C=C2O1
Molecular Weight (g/mol) 246.26
Formula Weight 246.26
Percent Purity 99.11%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.