missing translation for 'onlineSavingsMsg'
Learn More

(S)-Mapracorat, MedChemExpress

Product Code. 16498807
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498807 5 mg 5mg
16456218 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16498807 Supplier Medchem Express Supplier No. HY14864A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (S)-Mapracorat is a selective and less active glucocorticoid receptor agonist.

(S)-Mapracorat is a selective and less active glucocorticoid receptor agonist.

Specifications

Chemical Name or Material (S)-Mapracorat
CAS 887375-15-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C25H26F4N2O2
Quantity 5 mg
Synonym (S)-ZK-245186 (S)-BOL-303242X
Solubility Information DMSO : ≥ 50.6 mg/mL (109.41 mM)
SMILES CC(C)(C1=C(OCC2)C2=CC(F)=C1)C[C@](O)(CNC3=C4C=CC(C)=NC4=CC=C3)C(F)(F)F
Molecular Weight (g/mol) 462.48
Formula Weight 462.48
Percent Purity 99.03%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.