missing translation for 'onlineSavingsMsg'
Learn More

(S)-Linagliptin, TRC

Product Code. 30008433
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008433 10 mg 10mg
30007698 100 mg 100mg
30012521 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008433 Supplier Toronto Research Chemicals Supplier No. TRCL46589510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 668270-11-9
Molecular Formula C25H28N8O2
Molecular Weight (g/mol) 472.54
Synonym 8-[(3S)-3-Amino-1-piperidinyl]-7-(2-butyn-1-yl)-3,7-dihydro-3-methyl-1-[(4-methyl-2-quinazolinyl)methyl]-1H-purine-2,6-dione
IUPAC Name 8-[(3S)-3-aminopiperidin-1-yl]-7-but-2-ynyl-3-methyl-1-[(4-methylquinazolin-2-yl)methyl]purine-2,6-dione
SMILES O=C(N(C)C(N=C(N1C[C@@H](N)CCC1)N2CC#CC)=C2C3=O)N3CC4=NC5=CC=CC=C5C(C)=N4

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C25H28N8O2/c1-4-5-13-32-21-22(29-24(32)31-12-8-9-17(26)14-31)30(3)25(35)33(23(21)34)15-20-27-16(2)18-10-6-7-11-19(18)28-20/h6-7,10-11,17H,8-9,12-15,26H2,1-3H3/t17-/m0/s1
Formula Weight 472.2335
Percent Purity >95%
Chemical Name or Material (S)-Linagliptin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.