missing translation for 'onlineSavingsMsg'
Learn More

(S)-Isotimolol, TRC

Product Code. 30389174
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389174 100 mg 100mg
30407693 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30389174 Supplier Toronto Research Chemicals Supplier No. TRCI918000100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 158636-96-5
Molecular Formula C13 H24 N4 O3 S
Molecular Weight (g/mol) 316.42
Synonym (S)-Isotimolol, Timolol Maleate Impurity B as S-Enantiomer, Timolol Impurity B as S-Enantiomer, Timolol related compound B as S-Enantiomer, (2S)-3-[(1,1-dimethylethyl)amino]-2-[[4-(morpholin-4-yl)-1,2,5-thiadiazol-3-yl]oxy]propan-1-ol
IUPAC Name (2S)-3-(tert-butylamino)-2-[(4-morpholin-4-yl-1,2,5-thiadiazol-3-yl)oxy]propan-1-ol
SMILES CC(C)(C)NC[C@@H](CO)Oc1nsnc1N2CCOCC2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C13H24N4O3S/c1-13(2,3)14-8-10(9-18)20-12-11(15-21-16-12)17-4-6-19-7-5-17/h10,14,18H,4-9H2,1-3H3/t10-/m0/s1
Formula Weight 316.1569
Chemical Name or Material (S)-Isotimolol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.