missing translation for 'onlineSavingsMsg'
Learn More

(S)-Isotimolol, Mikromol™

Product Code. 17909939
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17909939 100 mg 100mg
17959609 25 mg 25mg
30464190 4 x 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17909939 Supplier Mikromol Supplier No. MM0440.02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(S)-Isotimolol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-Isotimolol
CAS 158636-96-5
Analyte or Component Names (S)-Isotimolol
Shipping Condition Room Temperature
Molecular Formula C13 H24 N4 O3 S
Certifications/Compliance ISO 17025
Synonym (S)-Isotimolol, Timolol Maleate Impurity B as S-Enantiomer, Timolol Impurity B as S-Enantiomer, Timolol related compound B as S-Enantiomer, (2S)-3-[(1,1-dimethylethyl)amino]-2-[[4-(morpholin-4-yl)-1,2,5-thiadiazol-3-yl]oxy]propan-1-ol
InChI Formula InChI=1S/C13H24N4O3S/c1-13(2,3)14-8-10(9-18)20-12-11(15-21-16-12)17-4-6-19-7-5-17/h10,14,18H,4-9H2,1-3H3/t10-/m0/s1
SMILES CC(C)(C)NC[C@@H](CO)Oc1nsnc1N2CCOCC2
IUPAC Name (2S)-3-(tert-butylamino)-2-[(4-morpholin-4-yl-1,2,5-thiadiazol-3-yl)oxy]propan-1-ol
Molecular Weight (g/mol) 316.42
Quantity 100 mg
Formula Weight 316.1569 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.