missing translation for 'onlineSavingsMsg'
Learn More

(S)-Isopropyl-2-aminopropanoate Hydrochloride, TRC

Product Code. 30398475
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398475 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30398475 Supplier Toronto Research Chemicals Supplier No. TRCI8222201G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-Isopropyl-2-aminopropanoate Hydrochloride
CAS 62062-65-1
Molecular Formula C6 H13 N O2 . H Cl
Quantity 1 g
Synonym L-Alanine Isopropyl Ester Hydrochloride, Isopropyl L-Alaninate Hydrochloride, L-Alanine 2-Propyl Ester Hydrochloride, L-Alanine Isopropyl Ester Hydrochloride, O-Isopropyl-L-alanine Hydrochloride
InChI Formula InChI=1S/C6H13NO2.ClH/c1-4(2)9-6(8)5(3)7;/h4-5H,7H2,1-3H3;1H/t5-;/m0./s1
SMILES Cl.CC(C)OC(=O)[C@H](C)N
IUPAC Name propan-2-yl (2S)-2-aminopropanoate;hydrochloride
Molecular Weight (g/mol) 167.634
Formula Weight 167.071
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.