missing translation for 'onlineSavingsMsg'
Learn More

(S)-Imazamox, TRC

Product Code. 30397070
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397070 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30397070 Supplier Toronto Research Chemicals Supplier No. TRCI2685555MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-Imazamox
CAS 355838-61-8
Molecular Formula C15H19N3O4
Quantity 5 mg
Synonym 5-Methoxymethyl-2-((4S)-4-isopropyl-4-methyl-5-oxo-2-imidazolin-2-yl) Nicotinic Acid, 2-[(4S)-4,5-Dihydro-4-methyl-4-(1-methylethyl)-5-oxo-1H-imidazol-2-yl]-5-(methoxymethyl)-3-pyridinecarboxylic Acid
InChI Formula InChI=1S/C15H19N3O4/c1-8(2)15(3)14(21)17-12(18-15)11-10(13(19)20)5-9(6-16-11)7-22-4/h5-6,8H,7H2,1-4H3,(H,19,20)(H,17,18,21)/t15-/m0/s1
SMILES COCc1cnc(C2=NC(=O)[C@@](C)(N2)C(C)C)c(c1)C(=O)O
IUPAC Name 5-(methoxymethyl)-2-[(5S)-5-methyl-4-oxo-5-propan-2-yl-1H-imidazol-2-yl]pyridine-3-carboxylic acid
Molecular Weight (g/mol) 305.33
Formula Weight 305.1376
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.