missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-Ibuprofen, TRC

Product Code. 17970335
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17970335 500 mg 500mg
17931605 100 mg 100mg
17934465 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17970335 Supplier Toronto Research Chemicals Supplier No. TRCI140010500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51146-56-6
Molecular Formula C13 H18 O2
Molecular Weight (g/mol) 206.28
Synonym Benzeneacetic acid, α-methyl-4-(2-methylpropyl)-, (αS)-, Benzeneacetic acid, α-methyl-4-(2-methylpropyl)-, (S)-, (+)-(S)-Ibuprofen, (+)-(S)-p-Isobutylhydratropic acid, (+)-Ibuprofen, (+)-Ibuprophen, (+)-α-Methyl-4-(2-methylpropyl)benzeneacetic acid, (S)-(+)-2-(4-Isobutylphenyl)propionic acid, (S)-(+)-4-Isobutyl-α-methylphenylacetic acid, (S)-(+)-Ibuprofen, (S)-(+)-p-Isobutylhydratropic acid, (S)-2-(4-Isobutylphenyl)propanoic acid, (S)-2-(4-Isobutylphenyl)propionic acid, (S)-2-(p-Isobutylphenyl)propionic acid, (S)-Ibuprofen, (αS)-α-Methyl-4-(2-methylpropyl)benzeneacetic acid, Dexibuprofen, Nyfen, S-Optifen, Seractil, Ultraprofen, d-Ibuprofen
IUPAC Name (2S)-2-[4-(2-methylpropyl)phenyl]propanoic acid
SMILES CC(C)Cc1ccc(cc1)[C@H](C)C(=O)O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C13H18O2/c1-9(2)8-11-4-6-12(7-5-11)10(3)13(14)15/h4-7,9-10H,8H2,1-3H3,(H,14,15)/t10-/m0/s1
Formula Weight 206.1307
Percent Purity >95%
Chemical Name or Material (S)-(+)-Ibuprofen
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.