missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-gamma-Hydroxymethyl-gamma-butyrolactone, 97%

Product Code. 10279793 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10279793 250 mg Glass Bottle 250mg
10643162 1 g Glass Bottle 1g
11632769 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10279793 Supplier Thermo Scientific Acros Supplier No. 337642500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 32780-06-6 | C5H8O3 | 116.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32780-06-6
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.12
MDL Number MFCD00066224
InChI Key NSISJFFVIMQBRN-BYPYZUCNSA-N
Synonym s-5-hydroxymethyl dihydrofuran-2 3h-one, s-+-dihydro-5-hydroxymethyl-2 3h-furanone, s-+-dihydro-5-hydroxymethyl furanone, s-+-gamma-hydroxymethyl-gamma-butyrolactone, 5s-5-hydroxymethyl oxolan-2-one, s-5-hydroxymethyldihydrofuran-2-one, s-5-hydroxymethyl-dihydrofuran-2 3h-one, 2 3h-furanone, dihydro-5-hydroxymethyl-, 5s, s-4-hydroxymethyl butyrolactone, s-+-2',3'-dideoxyribonolactone
PubChem CID 2733813
IUPAC Name (5S)-5-(hydroxymethyl)oxolan-2-one
SMILES C1CC(=O)OC1CO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Density 1.2300g/mL
Boiling Point 110°C to 115°C (2.0 mmHg)
Flash Point >110°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4680 to 1.4720
Quantity 250 mg
Specific Gravity 1.23
Specific Rotation Condition +33.5° (20°C 589nm,c=3,EtOH)
Specific Rotation + 33.50
Solubility Information Solubility in water: miscible.
Formula Weight 116.12
Percent Purity 97%
Chemical Name or Material (S)-(+)-γ-Hydroxymethyl-γ-butyrolactone
Show More Show Less
compliance-icons
Product Identifier
  • (S)-(+)-y-Hydroxymethyl-y-butyrolactone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.