missing translation for 'onlineSavingsMsg'
Learn More

(S)-Homophenylalaninol-d2, TRC

Product Code. 30396323
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396323 1 g 1g
30406730 50 mg 50mg
30402142 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30396323 Supplier Toronto Research Chemicals Supplier No. TRCH6005121G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-Homophenylalaninol-d2
Molecular Formula C10 D2 H13 N O
Quantity 1 g
Synonym (2S)-2-Amino-4-phenyl-1-butanol-d2, (S)-beta-Aminobenzenebutanol-d2, L-2-Amino-4-phenyl-1-butanol-d2, (betaS)-beta-Aminobenzenebutanol-d2
InChI Formula InChI=1S/C10H15NO/c11-10(8-12)7-6-9-4-2-1-3-5-9/h1-5,10,12H,6-8,11H2/t10-/m0/s1/i8D2
SMILES [2H]C([2H])(O)[C@@H](N)CCc1ccccc1
IUPAC Name (2S)-2-amino-1,1-dideuterio-4-phenylbutan-1-ol
Molecular Weight (g/mol) 167.244
Formula Weight 167.128
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.