Learn More
Tocris Bioscience™ (S)-(-)-HA-966
NMDA antagonist/partial agonist
Specifications
Assay Percent Range | >98% |
---|---|
Chemical Name or Material | (S)-(-)-3-Amino-1-hydroxypyrrolidin-2-one |
Description
Possesses potent sedative and ataxic action, probably through disruption of striatal dopaminergic mechanisms.R-enantiomer also available.
Specifications
>98% | |
(S)-(-)-3-Amino-1-hydroxypyrrolidin-2-one |
Safety and Handling
Recommended Storage : Store at Room Temperature
For Research Use Only