missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Glycidyl trityl ether, 98%, ee 98%, Thermo Scientific™

Product Code. 15455747
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15455747 1 g 1g
15465747 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15455747 Supplier Thermo Scientific Alfa Aesar Supplier No. H32961.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129940-50-7
Molecular Formula C22H20O2
Molecular Weight (g/mol) 316.4
MDL Number MFCD00723373
InChI Key XFSXUCMYFWZRAF-NRFANRHFSA-N
Synonym s---trityl glycidyl ether, s-2-trityloxy methyl oxirane, s-trityl glycidyl ether, s---glycidyl trityl ether, s-glycidyl trityl ether, 2s-2-trityloxymethyl oxirane, s-tritylglycidylether, s-2-triphenylmethoxymethyl oxirane, s---glycidyl triphenylmethyl ether, s-glycidyl triphenylmethyl ether
PubChem CID 7168113
IUPAC Name (2S)-2-(trityloxymethyl)oxirane
SMILES C1C(O1)COC(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 99°C to 102°C
Quantity 1 g
Optical Rotation −10.5° (c=1 in Chloroform)
Formula Weight 316.39
Percent Purity 98%
Assay ee 98%
Chemical Name or Material (S)-(-)-Glycidyl trityl ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.