missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Glycidol, 99+%, ee 99+%

Product Code. 15480229
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15480229 5 g 5g
15470229 1 g 1g
15490229 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15480229 Supplier Thermo Scientific Alfa Aesar Supplier No. H63502.06
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 60456-23-7 | C3H6O2 | 74.079 g/mol

(S)-(-)-Glycidol is employed in the synthesis of chiral (E)-allylic alcohol side-chains for introduction into prostacyclin and prostaglandin frameworks by cross-metathesis. Also used as a starting reagent to synthesize non-peptidic HIV protease inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-Glycidol is employed in the synthesis of chiral (E)-allylic alcohol side-chains for introduction into prostacyclin and prostaglandin frameworks by cross-metathesis. Also used as a starting reagent to synthesize non-peptidic HIV protease inhibitors.

Solubility
Soluble in chloroform, DMSO and methanol.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60456-23-7
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.079
MDL Number MFCD00074874
InChI Key CTKINSOISVBQLD-VKHMYHEASA-N
Synonym s-glycidol, s-oxiranemethanol, s---glycidol, s-oxiran-2-ylmethanol, s---2,3-epoxy-1-propanol, 2s-oxiran-2-ylmethanol, s-+-glycidol, 2s-oxiran-2-yl methanol, oxiranemethanol, 2s
PubChem CID 6973630
ChEBI CHEBI:38690
IUPAC Name [(2S)-oxiran-2-yl]methanol
SMILES C1C(O1)CO

Specifications

Color Yellow
Density 1.116 g/mL
Boiling Point 66°C to 67°C (19 mmHg)
Flash Point 66°C (151°F)
Refractive Index 1.433
Quantity 5 g
UN Number UN2810
Beilstein 79783
Solubility Information Soluble in chloroform,DMSO and methanol.
Formula Weight 74.08
Percent Purity ≥99%, ee
Physical Form Liquid
Chemical Name or Material (S)-(-)-Glycidol, ee ≥99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.