missing translation for 'onlineSavingsMsg'
Learn More

(S)-Flurbiprofen (S)-(-)-a-Methylbenzylamine Salt, TRC

Product Code. 30396697
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396697 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30396697 Supplier Toronto Research Chemicals Supplier No. TRCF5987162.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-Flurbiprofen (S)-(-)-alpha-Methylbenzylamine Salt
CAS 76319-57-8
Molecular Formula C15H13FO2 C8H11N
Quantity 2.5 g
Synonym (2S)-2-(2-Fluoro-1,1'-biphenyl-4-yl)propanoic Acid (S)-(-)-alpha-Methylbenzylamine Salt, (2S)-2-(2-Fluoro-4-biphenyl)propanoic Acid (S)-(-)-alpha-Methylbenzylamine Salt
InChI Formula InChI=1S/C15H13FO2.C8H11N/c1-10(15(17)18)12-7-8-13(14(16)9-12)11-5-3-2-4-6-11;1-7(9)8-5-3-2-4-6-8/h2-10H,1H3,(H,17,18);2-7H,9H2,1H3/t10-;7-/m00/s1
SMILES C[C@H](N)c1ccccc1.C[C@H](C(=O)O)c2ccc(c(F)c2)c3ccccc3
IUPAC Name (2S)-2-(3-fluoro-4-phenylphenyl)propanoic acid;(1S)-1-phenylethanamine
Molecular Weight (g/mol) 365.4
Formula Weight 365.1791
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.