missing translation for 'onlineSavingsMsg'
Learn More

(S)-Fluoxetine Hydrochloride, TRC

Product Code. 30012274
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012274 1 mg 1mg
30010114 5 mg 5mg
30005507 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012274 Supplier Toronto Research Chemicals Supplier No. TRCF5970951MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114247-06-2
Molecular Formula C17H19ClF3NO
Molecular Weight (g/mol) 345.79
Synonym Benzenepropanamine, N-methyl-γ-[4-(trifluoromethyl)phenoxy]-, hydrochloride (1:1), (γS)-, Benzenepropanamine, N-methyl-γ-[4-(trifluoromethyl)phenoxy]-, hydrochloride, (S)-, Benzenepropanamine, N-methyl-γ-[4-(trifluoromethyl)phenoxy]-, hydrochloride, (γS)- (9CI), (+)-(S)-Fluoxetine hydrochloride, (S)-Fluoxetine hydrochloride
SMILES Cl.CNCC[C@H](Oc1ccc(cc1)C(F)(F)F)c2ccccc2

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C17H18F3NO.ClH/c1-21-12-11-16(13-5-3-2-4-6-13)22-15-9-7-14(8-10-15)17(18,19)20;/h2-10,16,21H,11-12H2,1H3;1H/t16-;/m0./s1
Formula Weight 345.1107
Percent Purity >95%
Chemical Name or Material (S)-Fluoxetine Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.