missing translation for 'onlineSavingsMsg'
Learn More

S-Ethyl thioacetate, 98+%

Product Code. 11443444
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11443444 100 g 100g
11423444 5 g 5g
11433444 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11443444 Supplier Thermo Scientific Alfa Aesar Supplier No. A10680.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 625-60-5 | C4H8OS | 104.167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-60-5
Molecular Formula C4H8OS
Molecular Weight (g/mol) 104.167
MDL Number MFCD00015178
InChI Key APTGPWJUOYMUCE-UHFFFAOYSA-N
Synonym s-ethyl thioacetate, ethanethioic acid, ethyl ester, ethanethioic acid, s-ethyl ester, ethyl thiolacetate, ethyl ethanethioate, ethanethioic acid s-ethyl ester, s-ethyl thiolacetate, acetic acid, thio-, ethyl ester, thioethyl compound, acetic acid, thio-, s-ethyl ester
PubChem CID 61171
IUPAC Name S-ethyl ethanethioate
SMILES CCSC(=O)C

Specifications

Density 0.98
Boiling Point 116°C to 117°C
Flash Point 20°C (68°F)
Odor Stench
Refractive Index 1.46
Quantity 100 g
UN Number UN1993
Beilstein 1737643
Solubility Information Insoluble in water
Formula Weight 104.18
Percent Purity ≥98%
Chemical Name or Material S-Ethyl thioacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.