missing translation for 'onlineSavingsMsg'
Learn More

(S)-Equol, TRC

Product Code. 30136840
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136840 25 mg 25mg
17958784 5 mg 5mg
17918124 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136840 Supplier Toronto Research Chemicals Supplier No. TRCE59300125MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 531-95-3
Molecular Formula C15 H14 O3
Molecular Weight (g/mol) 242.27
Synonym 2H-1-Benzopyran-7-ol, 3,4-dihydro-3-(4-hydroxyphenyl)-, (3S)-, 2H-1-Benzopyran-7-ol, 3,4-dihydro-3-(4-hydroxyphenyl)-, (S)-, 4',7-Isoflavandiol (6CI,7CI,8CI), (3S)-3,4-Dihydro-3-(4-hydroxyphenyl)-2H-1-benzopyran-7-ol, (-)-(S)-Equol, (-)-Equol, (3S)-3-(4-Hydroxyphenyl)-3,4-dihydro-2H-chromen-7-ol, (3S)-Equol, (S)-(-)-4',7-Isoflavandiol, (S)-(-)-Equol, (S)-3-(4-Hydroxyphenyl)chroman-7-ol, (S)-Equol, 4',7-Dihydroxyisoflavan, AUS 131, Equol, Equol, (-)-, S-equol, SE 5OH
IUPAC Name (3S)-3-(4-hydroxyphenyl)-3,4-dihydro-2H-chromen-7-ol
SMILES Oc1ccc(cc1)[C@H]2COc3cc(O)ccc3C2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C15H14O3/c16-13-4-1-10(2-5-13)12-7-11-3-6-14(17)8-15(11)18-9-12/h1-6,8,12,16-17H,7,9H2/t12-/m1/s1
Formula Weight 242.0943
Percent Purity >95%
Chemical Name or Material (S)-Equol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.