missing translation for 'onlineSavingsMsg'
Learn More

(-)-(S)-Equol, MedChemExpress

Product Code. 16493088
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493088 10 mg 10mg
16479646 50 mg 50mg
16462616 25 mg 25mg
16445827 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16493088 Supplier Medchem Express Supplier No. HY100583
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress (-)-(S)-Equol is a high affinity ligand for estrogen receptor β with a Ki of 0.73 nM.

(-)-(S)-Equol is a high affinity ligand for estrogen receptor β with a Ki of 0.73 nM.

Specifications

Chemical Name or Material (-)-(S)-Equol
CAS 531-95-3
Color White
Purity Grade Notes Research
Molecular Formula C15H14O3
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (412.76 mM; Need ultrasonic and warming)
SMILES OC1=CC=C2C[C@@H](C3=CC=C(O)C=C3)COC2=C1
Molecular Weight (g/mol) 242.27
Formula Weight 242.27
Percent Purity 98.78%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.