missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-Epichlorohydrin, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | (S)-Epichlorohydrin |
| CAS | 67843-74-7 |
| Molecular Formula | C3 H5 Cl O |
| Quantity | 1 g |
| Synonym | Oxirane, 2-(chloromethyl)-, (2S)-, Oxirane, (chloromethyl)-, (2S)- (9CI), Oxirane, (chloromethyl)-, (S)-, (2S)-2-(Chloromethyl)oxirane, (+)-2-(Chloromethyl)oxirane, (+)-Epichlorohydrin, (2S)-2-(Chloromethyl)oxirane, (S)-(+)-Epichlorohydrin, (S)-(Chloromethyl)oxirane, (S)-1-Chloro-2,3-epoxypropane, (S)-2-Chloromethyloxirane, (S)-Epichlorhydrin, (S)-Epichlorohydrin, 2(S)-Epichlorohydrin |
| InChI Formula | InChI=1S/C3H5ClO/c4-1-3-2-5-3/h3H,1-2H2/t3-/m1/s1 |
| SMILES | ClC[C@@H]1CO1 |
| IUPAC Name | (2S)-2-(chloromethyl)oxirane |
| Molecular Weight (g/mol) | 92.52 |
| Formula Weight | 92.0029 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction