missing translation for 'onlineSavingsMsg'
Learn More

(S)-Duloxetine Succinamide, TRC

Product Code. 30385966
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385966 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385966 Supplier Toronto Research Chemicals Supplier No. TRCD72101525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-4-(Methyl[3-(naphthalen-1-yloxy)-3-(thiophen-2-yl)propyl]amino)-4-oxobutanoic acid
CAS 199191-66-7
Purity Grade Notes HPLC
Molecular Formula C22 H23 N O4 S
Quantity 25 mg
Synonym 4-[methyl[(3S)-3-(1-naphthalenyloxy)-3-(2-thienyl)propyl]amino]-4-oxo-butanoic acid, (S)-Duloxetine Succinamide, Duloxetine Related Compound H (USP)
InChI Formula InChI=1S/C22H23NO4S/c1-23(21(24)11-12-22(25)26)14-13-19(20-10-5-15-28-20)27-18-9-4-7-16-6-2-3-8-17(16)18/h2-10,15,19H,11-14H2,1H3,(H,25,26)/t19-/m0/s1
SMILES CN(CC[C@H](Oc1cccc2ccccc12)c3cccs3)C(=O)CCC(=O)O
IUPAC Name 4-[methyl-[(3S)-3-naphthalen-1-yloxy-3-thiophen-2-ylpropyl]amino]-4-oxobutanoic acid
Molecular Weight (g/mol) 397.49
Formula Weight 397.1348
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.