missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-Duloxetine Succinamide, TRC
Specifications
Specifications
| Chemical Name or Material | (S)-4-(Methyl[3-(naphthalen-1-yloxy)-3-(thiophen-2-yl)propyl]amino)-4-oxobutanoic acid |
| CAS | 199191-66-7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C22 H23 N O4 S |
| Quantity | 50 mg |
| Synonym | 4-[methyl[(3S)-3-(1-naphthalenyloxy)-3-(2-thienyl)propyl]amino]-4-oxo-butanoic acid, (S)-Duloxetine Succinamide, Duloxetine Related Compound H (USP) |
| InChI Formula | InChI=1S/C22H23NO4S/c1-23(21(24)11-12-22(25)26)14-13-19(20-10-5-15-28-20)27-18-9-4-7-16-6-2-3-8-17(16)18/h2-10,15,19H,11-14H2,1H3,(H,25,26)/t19-/m0/s1 |
| SMILES | CN(CC[C@H](Oc1cccc2ccccc12)c3cccs3)C(=O)CCC(=O)O |
| IUPAC Name | 4-[methyl-[(3S)-3-naphthalen-1-yloxy-3-thiophen-2-ylpropyl]amino]-4-oxobutanoic acid |
| Molecular Weight (g/mol) | 397.49 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?