missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ (S)-Duloxetine hydrochloride

Product Code. 30001596
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001596 50 mg 50mg
30001576 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001596 Supplier R&D Systems™ Supplier No. 4798/50
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent 5-HT and NA reuptake inhibitor (SNRI); also blocks dopamine reuptake

High affinity, competitive 5-HT and noradrenaline (NA) re-uptake inhibitor (Ki values are 8.5 and 45 nM for 5-HT and NA reuptake respectively in cortical synaptosomes; IC50 values are 28 and 46 nM for 5-HT and NA reuptake respectively in rat hippocampal slices). Also blocks dopamine reuptake (Ki = 300 nM in striatal synaptosomes). Exhibits antidepressant and anxiolytic effects. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136434-34-9
Molecular Formula C18H20ClNOS

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 333.88
Purity >99%
Molecular Weight (g/mol) 333.87
Chemical Name or Material (S)-Duloxetine hydrochloride
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.