missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ (S)-(+)-Dimethindene maleate
GREENER_CHOICE

Product Code. 18712634
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18712634

Brand: Tocris Bioscience™ 1425/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

M2-selective antagonist

(S)-(+)-Dimethindene maleate is an enantiomer that is a subtype-selective M2 muscarinic receptor antagonist (pKi values are 7.08, 7.78, 6.70 and 7.00 for M1, M2, M3 and M4 receptors respectively). Also H1 histamine receptor antagonist (pKi = 7.48). Allows formation of extended pluripotent stem (EPS) cells in combination with CHIR 99021 (Cat.No. 4423), minocycline hydrochloride (Cat.No. 3268) and human leukemia inhibitory factor.

For more information about how Dimethindene maleate may be used, see our protocol: Generation and Propagation of EPS cells (LCDM Cocktail) .

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136152-65-3
Molecular Formula C24H28N2O4

Specifications

Quantity 50 mg
Formula Weight 408.5
Purity >99%
Molecular Weight (g/mol) 408.498
Chemical Name or Material (S)-(+)-Dimethindene maleate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.