missing translation for 'onlineSavingsMsg'
Learn More

(+)-(S)-Dihydrokavain, TRC

Product Code. 30375040
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30375040 5 mg
30384945 1 mg
30384727 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30375040 Supplier Toronto Research Chemicals Supplier No. TRCD4506205MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 587-63-3
Molecular Formula C14 H16 O3
Molecular Weight (g/mol) 232.28
Synonym 2H-Pyran-2-one, 5,6-dihydro-4-methoxy-6-(2-phenylethyl)-, (6S)- (9CI), 2H-Pyran-2-one, 5,6-dihydro-4-methoxy-6-(2-phenylethyl)-, (S)-, 2H-Pyran-2-one, 5,6-dihydro-4-methoxy-6-phenethyl- (8CI), (+)-7,8-Dihydrokavain, (+)-Dihydrokawain, 7,8-Dihydrokavain, 7,8-Dihydrokawain, Dihydrokavain, Dihydrokawain, Kawain, dihydro-, Marindinin, NSC 112163, (+)-Dihydrokavain
IUPAC Name (2S)-4-methoxy-2-(2-phenylethyl)-2,3-dihydropyran-6-one
SMILES COC1=CC(=O)O[C@@H](CCc2ccccc2)C1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C14H16O3/c1-16-13-9-12(17-14(15)10-13)8-7-11-5-3-2-4-6-11/h2-6,10,12H,7-9H2,1H3/t12-/m0/s1
Formula Weight 232.1099
Chemical Name or Material Dihydrokavain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.