missing translation for 'onlineSavingsMsg'
Learn More

(S)-Desmethyl Citalopram Ethanedioate, TRC

Product Code. 30365786
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30365786 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30365786 Supplier Toronto Research Chemicals Supplier No. TRCD29151510MG
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trityl Olmesartan
CAS 852172-06-6
Molecular Formula C19 H19 F N2 O . C2 H2 O4
Quantity 10 mg
Synonym 5-Isobenzofurancarbonitrile, 1-(4-fluorophenyl)-1,3-dihydro-1-[3-(methylamino)propyl]-, (1S)-, ethanedioate (1:1) (9CI), (1S)-1-(4-Fluorophenyl)-1,3-dihydro-1-[3-(methylamino)propyl]-5-isobenzofurancarbonitrile oxalate, (S)-1-(3-Methylaminopropyl)-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-5-carbonitrile oxalate, (S)-Desmethylcitalopram oxalate
InChI Formula InChI=1S/C19H19FN2O.C2H2O4/c1-22-10-2-9-19(16-4-6-17(20)7-5-16)18-8-3-14(12-21)11-15(18)13-23-19;3-1(4)2(5)6/h3-8,11,22H,2,9-10,13H2,1H3;(H,3,4)(H,5,6)/t19-;/m0./s1
SMILES CNCCC[C@]1(OCc2cc(ccc12)C#N)c3ccc(F)cc3.OC(=O)C(=O)O
IUPAC Name (1S)-1-(4-fluorophenyl)-1-[3-(methylamino)propyl]-3H-2-benzofuran-5-carbonitrile;oxalic acid
Molecular Weight (g/mol) 400.4
Formula Weight 400.1434
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.