Learn More
S-Curve™ Two Compartment Packaged Glove Dispenser
Product Code. 16480905
Encompass_Preferred
Change view
Click to view available options
Material:
PETG
Unit Size:
Each
Product Code. Material Unit Size
16480905 PETG Each
1 options
Product Code. 16480905 Supplier S-Curve™ Supplier No. 19999451
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Specially designed for sterile or packaged gloves, with two individual compartments that dispense gloves in a first-in first-out fashion.
  • Specially designed for sterile or pair-packaged gloves
  • Gloves dispense in a first-in, first-out fashion
  • Single, sloped lid with a full-length hinge to maintain HEPA flow
  • Can be wall-mounted with heavy-duty mounting bracket (included)
  • Flame-treated edges for a smooth finish
Specifications
Product Type Glove Dispenser
Color Black
Dimensions (D x W x H) Interior 12 x 14 x 16 in.
Holds Sterile Pair-Packaged Gloves
Material PETG
Mounting Keyhole Slots
No. of Compartments 2
Product Style Top Loading
Includes Heavy-duty wall mounting bracket
Thickness (English) 0.25 in.