missing translation for 'onlineSavingsMsg'
Learn More

S-(-)-Cotinine, TRC

Product Code. 30135727
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135727 1 g 1g
17991375 250 mg 250mg
17913405 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135727 Supplier Toronto Research Chemicals Supplier No. TRCC7250001G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 486-56-6
Molecular Formula C10 H12 N2 O
Molecular Weight (g/mol) 176.22
Synonym 2-Pyrrolidinone, 1-methyl-5-(3-pyridinyl)-, (5S)-, 2-Pyrrolidinone, 1-methyl-5-(3-pyridinyl)-, (S)-, Cotinine (6CI,7CI,8CI), (5S)-1-Methyl-5-(3-pyridinyl)-2-pyrrolidinone, (-)-Cotinine, (S)-Cotinine, NIH 10498, S-(-)-Cotinine, (5S)-1-Methyl-5-(pyridin-3-yl)pyrrolidin-2-one, Cotinine
IUPAC Name (5S)-1-methyl-5-pyridin-3-ylpyrrolidin-2-one
SMILES CN1[C@@H](CCC1=O)c2cccnc2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H12N2O/c1-12-9(4-5-10(12)13)8-3-2-6-11-7-8/h2-3,6-7,9H,4-5H2,1H3/t9-/m0/s1
Formula Weight 176.095
Percent Purity >95%
Chemical Name or Material Cotinine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.