missing translation for 'onlineSavingsMsg'
Learn More

(S)-Cotinine N-Oxide, TRC

Product Code. 30137289
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137289 100 mg 100mg
30009205 10 mg 10mg
30012583 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137289 Supplier Toronto Research Chemicals Supplier No. TRCC725200100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36508-80-2
Molecular Formula C10 H12 N2 O2
Molecular Weight (g/mol) 192.21
Synonym 2-Pyrrolidinone, 1-methyl-5-(1-oxido-3-pyridinyl)-, (5S)- (9CI, ACI), (5S)-1-Methyl-5-(1-oxido-3-pyridinyl)-2-pyrrolidinone (ACI), 2-Pyrrolidinone, 1-methyl-5-(3-pyridinyl)-, N-oxide, (S)- (ZCI), (5S)-1-Methyl-5-(1-oxidopyridin-1-ium-3-yl)pyrrolidin-2-one, (5S)-Cotinine N-oxide, 2-Pyrrolidinone, 1-methyl-5-(1-oxido-3-pyridinyl)-, (S)-, 3-[(2S)-1-Methyl-5-oxopyrrolidin-2-yl]pyridin-1-ium-1-olate, Cotinine N-oxide, (S)-Cotinine N-oxide
IUPAC Name (5S)-1-methyl-5-(1-oxidopyridin-1-ium-3-yl)pyrrolidin-2-one
SMILES CN1[C@@H](CCC1=O)c2ccc[n+]([O-])c2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H12N2O2/c1-11-9(4-5-10(11)13)8-3-2-6-12(14)7-8/h2-3,6-7,9H,4-5H2,1H3/t9-/m0/s1
Formula Weight 192.0899
Chemical Name or Material (S)-Cotinine N-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.