missing translation for 'onlineSavingsMsg'
Learn More

S-(-)-Cotinine (1.0 mg/mL in Methanol), TRC

Product Code. 30399571
Change view
Click to view available options
Quantity:
10 x 1 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399571 10 x 1 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30399571 Supplier Toronto Research Chemicals Supplier No. TRCKIT054010X1ML
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 486-56-6
Molecular Formula C10 H12 N2 O
Quantity 10 x 1 mL
Synonym 2-Pyrrolidinone, 1-methyl-5-(3-pyridinyl)-, (5S)-, 2-Pyrrolidinone, 1-methyl-5-(3-pyridinyl)-, (S)-, Cotinine (6CI,7CI,8CI), (5S)-1-Methyl-5-(3-pyridinyl)-2-pyrrolidinone, (-)-Cotinine, (S)-Cotinine, NIH 10498, S-(-)-Cotinine, (5S)-1-Methyl-5-(pyridin-3-yl)pyrrolidin-2-one, Cotinine
InChI Formula InChI=1S/C10H12N2O/c1-12-9(4-5-10(12)13)8-3-2-6-11-7-8/h2-3,6-7,9H,4-5H2,1H3/t9-/m0/s1
SMILES CN1[C@@H](CCC1=O)c2cccnc2
IUPAC Name (5S)-1-methyl-5-pyridin-3-ylpyrrolidin-2-one
Molecular Weight (g/mol) 176.22
Formula Weight 176.095
Concentration or Composition (by Analyte or Components) Cotinine 1 mg/mL
Chemical Name or Material Cotinine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.