missing translation for 'onlineSavingsMsg'
Learn More

(S)-cis-Verbenol, 97%, sum of isomers

Product Code. 10224634 Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10224634

Brand: Thermo Scientific Acros 347330050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18881-04-4 | C10H16O | 152.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18881-04-4
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00065444
InChI Key WONIGEXYPVIKFS-YIZRAAEISA-N
Synonym s-cis-verbenol, verbenol, s-cis, unii-xr9t57f48t, cis-verbenol, bicyclo 3.1.1 hept-3-en-2-ol, 4,6,6-trimethyl-, 1s,2s,5s, 1s-cis-verbenol, ccris 5289, ?-cis-verbenol
PubChem CID 87839
IUPAC Name (1S,2S,5S)-4,6,6-trimethylbicyclo[3.1.1]hept-3-en-2-ol
SMILES CC1=CC(C2CC1C2(C)C)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 62°C to 68°C
Color Bordeaux to Brown-Red
Density 85g/mL
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (sum of isomers) (GC)
Packaging Glass bottle
Refractive Index 1.49
Quantity 5 g
Specific Rotation Condition + 10.00 (20.00°C c=2,C2H5OH)
Specific Rotation + 10
Solubility Information Solubility in water: insoluble
Formula Weight 152.24
Percent Purity 97%
Physical Form Powder
Chemical Name or Material (S)-cis-Verbenol, Sum of isomers
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.