missing translation for 'onlineSavingsMsg'
Learn More

(S)-Carisbamate, TRC

Product Code. 30009871
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009871 5 mg 5mg
30003349 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009871 Supplier Toronto Research Chemicals Supplier No. TRCC1835105MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 194085-75-1
Molecular Formula C9 H10 Cl N O3
Molecular Weight (g/mol) 215.63
Synonym 1,2-Ethanediol, 1-(2-chlorophenyl)-, 2-carbamate, (1S)-, 1,2-Ethanediol, 1-(2-chlorophenyl)-, 2-carbamate, (S)-, Carisbamate, Comfyde, JNJ 10234094, RWJ 333369, YKP 509, (S)-Carisbamate
IUPAC Name [(2S)-2-(2-chlorophenyl)-2-hydroxyethyl] carbamate
SMILES NC(=O)OC[C@@H](O)c1ccccc1Cl

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C9H10ClNO3/c10-7-4-2-1-3-6(7)8(12)5-14-9(11)13/h1-4,8,12H,5H2,(H2,11,13)/t8-/m1/s1
Formula Weight 215.0349
Chemical Name or Material (S)-Carisbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.