missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Bupivacaine, TRC

Product Code. 30010910
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010910 100 mg 100mg
30006802 10 mg 10mg
30003591 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010910 Supplier Toronto Research Chemicals Supplier No. TRCB689546100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27262-47-1
Molecular Formula C18 H28 N2 O
Molecular Weight (g/mol) 288.43
Synonym 2-Piperidinecarboxamide, 1-butyl-N-(2,6-dimethylphenyl)-, (2S)-, 2-Piperidinecarboxamide, 1-butyl-N-(2,6-dimethylphenyl)-, (S)-, 2',6'-Pipecoloxylidide, 1-butyl-, (-)- (8CI), (2S)-1-Butyl-N-(2,6-dimethylphenyl)-2-piperidinecarboxamide, (-)-(S)-Bupivacaine, (-)-Bupivacaine, (S)-(-)-Bupivacaine, (S)-Bupivacaine, Chirocain, L-(-)-Bupivacaine, Levobupivacaine
IUPAC Name (2S)-1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide
SMILES CCCCN1CCCC[C@H]1C(=O)Nc2c(C)cccc2C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C18H28N2O/c1-4-5-12-20-13-7-6-11-16(20)18(21)19-17-14(2)9-8-10-15(17)3/h8-10,16H,4-7,11-13H2,1-3H3,(H,19,21)/t16-/m0/s1
Formula Weight 288.2202
Chemical Name or Material (S)-(-)-Bupivacaine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.