missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-BINAP, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | (S)-BINAP |
| CAS | 76189-56-5 |
| Molecular Formula | C44H32P2 |
| Quantity | 1 g |
| Synonym | 1,1'-[(1S)-[1,1'-Binaphthalene]-2,2'-diyl]bis[1,1-diphenyl-phosphine, (1S)-[1,1'-Binaphthalene]-2,2'-diylbis[diphenylphosphine], (S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene, (S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl |
| InChI Formula | InChI=1S/C44H32P2/c1-5-19-35(20-6-1)45(36-21-7-2-8-22-36)41-31-29-33-17-13-15-27-39(33)43(41)44-40-28-16-14-18-34(40)30-32-42(44)46(37-23-9-3-10-24-37)38-25-11-4-12-26-38/h1-32H |
| SMILES | c1ccc(cc1)P(c2ccccc2)c3ccc4ccccc4c3c5c(ccc6ccccc56)P(c7ccccc7)c8ccccc8 |
| IUPAC Name | [1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane |
| Molecular Weight (g/mol) | 622.67 |
| Formula Weight | 622.1979 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction