missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-Benzyl Glycidyl Ether, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | (S)-(+)-Benzyl Glycidyl Ether |
| CAS | 16495-13-9 |
| Molecular Formula | C10H12O2 |
| Quantity | 1 g |
| Synonym | (2S)-2-[(Phenylmethoxy)methyl]oxirane, (2S)[(Phenylmethoxy)methyl]oxirane, , (S)-[(Phenylmethoxy)methyl]oxirane, (S)-1-(Benzyloxy)-2,3-epoxypropane, (+)-2-(Benzyloxymethyl)oxirane, (+)-Benzyl Glycidyl Ether, (+)-Glycidyl Benzyl Ether, (2S)-2-[(Benzyloxy)methyl]oxirane, (S)-(Benzyloxymethyl)oxirane, (S)-2-(Benzyloxymethyl)oxirane, (S)-Benzylglycidol, (S)-Glycidyl Benzyl Ether, (S)-O-Benzylglycidol, Benzyl (S)-Glycidyl Ether |
| InChI Formula | InChI=1S/C10H12O2/c1-2-4-9(5-3-1)6-11-7-10-8-12-10/h1-5,10H,6-8H2/t10-/m1/s1 |
| SMILES | C(OCc1ccccc1)[C@@H]2CO2 |
| IUPAC Name | (2S)-2-(phenylmethoxymethyl)oxirane |
| Molecular Weight (g/mol) | 164.2 |
| Formula Weight | 164.0837 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction