missing translation for 'onlineSavingsMsg'
Learn More

(S)-Benzyl 1-[(S)-2-benzyloxycarbonylamino-6-(2,2,2-trifluoroacetamido)hexanoyl]pyrrolidine-2-carboxylate, 95%, Thermo Scientific™

Product Code. 15405195
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15405195 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15405195 Supplier Thermo Scientific Alfa Aesar Supplier No. H34160.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 114753-64-9
Molecular Formula C28H32F3N3O6
MDL Number MFCD22380709
Quantity 1 g
Synonym s-benzyl 1-s-2-benzyloxycarbonylamino-6-2,2,2-trifluoroacetamido hexanoyl pyrrolidine-2-carboxylate, n2-phenylmethoxycarbonyl-n6-2,2,2-trifluoroacetyl-l-lysyl-l-proline phenylmethyl ester
InChI Key BWJDOHAQVMSNMG-NQCNTLBGSA-N
SMILES C1CC(N(C1)C(=O)C(CCCCNC(=O)C(F)(F)F)NC(=O)OCC2=CC=CC=C2)C(=O)OCC3=CC=CC=C3
IUPAC Name benzyl 1-[(2S)-2-(phenylmethoxycarbonylamino)-6-[(2,2,2-trifluoroacetyl)amino]hexanoyl]pyrrolidine-2-carboxylate
Molecular Weight (g/mol) 563.574
PubChem CID 73995171
Formula Weight 563.57
Percent Purity 95%
Chemical Name or Material (S)-Benzyl 1-[(S)-2-benzyloxycarbonylamino-6-(2,2,2-trifluoroacetamido)hexanoyl]pyrrolidine-2-carbox
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.