missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Bay-K-8644, MedChemExpress

Product Code. 16432437
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16432437

Brand: Medchem Express HY15124

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (S)-(-)-Bay-K-8644 is an agonist of L-type Ca2+ channel. (S)-(-)-Bay-K-8644 activates Ba2+ currents (IBa) (EC50=32 nM).

(S)-(-)-Bay-K-8644 is an agonist of L-type Ca2+ channel. (S)-(-)-Bay-K-8644 activates Ba2+ currents (IBa) (EC50=32 nM).

Specifications

Chemical Name or Material (S)-(-)-Bay-K-8644
CAS 98625-26-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C16H15F3N2O4
Quantity 50 mg
Solubility Information DMSO : 125 mg/mL (350.83 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES CC1=C([N+]([O-])=O)[C@@H](C2=C(C(F)(F)F)C=CC=C2)C(C(OC)=O)=C(C)N1
Molecular Weight (g/mol) 356.3
Formula Weight 356.3
Percent Purity 98.03%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.