missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Azetidine-2-carboxylic acid, 99%, Thermo Scientific Chemicals

Product Code. 11362197
Change view
Click to view available options
Quantity:
50 mg
250 mg
Unit Size:
250mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11362197 250 mg 250mg
11352197 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362197 Supplier Thermo Scientific Alfa Aesar Supplier No. L09668.MD
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2133-34-8 | Molecular Formula: C4H7NO2 | Molecular Weight: 101.105 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2133-34-8
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.105
MDL Number MFCD00005166
InChI Key IADUEWIQBXOCDZ-VKHMYHEASA-N
Synonym l-azetidine-2-carboxylic acid, s-azetidine-2-carboxylic acid, s---2-azetidinecarboxylic acid, 2s-azetidine-2-carboxylic acid, azetidyl-2-carboxylic acid, l-azetidine 2-carboxylic acid, s-2-azetidinecarboxylic acid, 2-azetidinecarboxylic acid, s, s---azetidine-2-carboxylic acid, 2-azetidinecarboxylic acid, l
PubChem CID 16486
ChEBI CHEBI:6198
IUPAC Name (2S)-azetidine-2-carboxylic acid
SMILES C1CNC1C(=O)O

Specifications

Melting Point ∽217°C (decomposition)
Quantity 250 mg
Beilstein 80678
Merck Index 14,908
Optical Rotation −123° (c=4 in H2O)
Formula Weight 101.11
Percent Purity 99%
Chemical Name or Material (S)-(-)-Azetidine-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.