Learn More
S-arrestin Antibody (S128), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-25161
 View more versions of this product
Product Code. 18747043
  • £377.00 / 0.10mL
Description
S-arrestin Monoclonal antibody specifically detects S-arrestin in Human, Mouse, Rat, Porcine, Bovine, Equine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunofluorescence
Western Blot 1:5000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Frozen
arrestin 1, RP47,48 kDa protein, S-antigen; retina and pineal gland (arrestin), S-arrestin, visual arrestin
Mouse, Rat, Pig, Bovine