missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S-arrestin Antibody (S128), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-25161SS
This item is not returnable.
View return policy
Description
S-arrestin Monoclonal antibody specifically detects S-arrestin in Human, Mouse, Rat, Porcine, Bovine, Equine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen).
Specifications
| S-arrestin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:5000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Frozen | |
| P10523 | |
| SAG | |
| Recombinant bovine S-arrestin with the first 20 amino acids of the C-terminus truncated [UniProt# P08168] | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Bovine, Mouse, Porcine and Rat. Other species have not been tested. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| S128 | |
| Unconjugated | |
| 50% PBS, 50% glycerol with 5mM Sodium Azide | |
| arrestin 1, RP47,48 kDa protein, S-antigen; retina and pineal gland (arrestin), S-arrestin, visual arrestin | |
| Mouse | |
| 48 kDa | |
| 0.025 mL | |
| Cell Biology, Cellular Markers, GPCR, Neuroscience, Neurotransmission, Sensory Systems, Signal Transduction, Vision | |
| 6295 | |
| Human, Mouse, Rat, Pig, Bovine, Equine, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction