missing translation for 'onlineSavingsMsg'
Learn More

S-arrestin Antibody (S128), Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18414457 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414457 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414457 Supplier Novus Biologicals Supplier No. NBP225161AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

S-arrestin Monoclonal antibody specifically detects S-arrestin in Human, Mouse, Rat, Porcine, Bovine, Equine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Specifications

Antigen S-arrestin
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone S128
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate
Gene Alias arrestin 1, RP47,48 kDa protein, S-antigen; retina and pineal gland (arrestin), S-arrestin, visual arrestin
Host Species Mouse
Immunogen Recombinant bovine S-arrestin with the first 20 amino acids of the C-terminus truncated [UniProt# P08168]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Neuroscience, Neurotransmission, Sensory Systems, Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 6295
Target Species Human, Mouse, Rat, Pig, Bovine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.