missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S-arrestin Antibody (PDS-1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB100-2385SS
This item is not returnable.
View return policy
Description
S-arrestin Monoclonal specifically detects S-arrestin in Human, Porcine, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| S-arrestin | |
| Monoclonal | |
| 0.77 mg/mL | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:100-1:200, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Frozen 1:100-1:200 | |
| SAG | |
| Porcine retinal S-arrestin [Swiss-Prot# P79260] | |
| 0.025 mL | |
| Cell Biology, Cellular Markers, GPCR, Neuroscience, Neurotransmission, Sensory Systems, Signal Transduction, Vision | |
| 6295 | |
| Human, Pig, Bovine | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| PDS-1 | |
| Unconjugated | |
| arrestin 1, RP47,48 kDa protein, S-antigen; retina and pineal gland (arrestin), S-arrestin, visual arrestin | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, cow, pig, and rat (weakly). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction