missing translation for 'onlineSavingsMsg'
Learn More

(S)-alpha-Methylphenylalanine hydrate, 98%, 98% ee, Thermo Scientific™

Product Code. 11995341 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11995341 100 mg 100mg
11905351 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11995341 Supplier Thermo Scientific Acros Supplier No. 275431000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 23239-35-2
Molecular Formula C10H13NO2·xH2O
Molecular Weight (g/mol) 179.22
InChI Key HYOWVAAEQCNGLE-JTQLQIEISA-N
Synonym alpha-methyl-l-phenylalanine, s-2-amino-2-methyl-3-phenylpropanoic acid, alpha-methyl-l-phe, alpha-methylphenylalanine, 2s-2-amino-2-methyl-3-phenylpropanoic acid, l-alpha-methylphenylalanine, alpha-benzyl-l-ala, s-2-amino-2-methyl-3-phenylpropionic acid, s-2-amimo-2-methyl-3-phenylpropanoic acid, s-alpha-methylphenylalanine
PubChem CID 2724754
IUPAC Name (2S)-2-amino-2-methyl-3-phenylpropanoic acid
SMILES CC(CC1=CC=CC=C1)(C(=O)O)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White
Enantiomeric Excess (HPLC)> 99.0%
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Quantity 100 mg
Specific Rotation − 18.00
Formula Weight 179.22
Percent Purity 98%,98% ee
Physical Form Fine Crystalline Powder
Chemical Name or Material (S)-α-Methylphenylalanine hydrate, 99%, (99+% e.e.)
Show More Show Less
Product Identifier
  • (S)-alpha-Methylphenylalanine hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.