missing translation for 'onlineSavingsMsg'
Learn More

(S)-alpha-Methylaspartic acid-4-tert-butyl ester, 98%, 98% ee, Thermo Scientific™

Product Code. 11985521 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 11985521

Brand: Thermo Scientific Acros 441171000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1217977-71-3
CAS Min % 97.5
CAS Max % 100.0
Melting Point 177°C to 178°C
Color White to Yellow
Enantiomeric Excess 98.0% min.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min
Molecular Formula C9H17NO4
MDL Number MFCD17018953
Quantity 100 mg
Specific Rotation 33
Synonym s-2-amino-4-tert-butoxy-2-methyl-4-oxobutanoic acid, s-alpha-methylaspartic acid-4-t-butyl ester, s-alpha-methylasparticacid-4-tert-butylester, s-alpha-methylaspartic acid-4-tert-butyl ester, h-alpha-me-l-asp tbu-oh, r-a-methylaspartic acid-4-tert-butyl ester, 2s-2-amino-4-tert-butoxy-2-methyl-4-oxobutanoic acid
InChI Key VIQZVYPYYUPDQY-UHFFFAOYNA-N
SMILES CC(C)(C)OC(=O)CC(C)(N)C(O)=O
IUPAC Name (2S)-2-amino-2-methyl-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid
Molecular Weight (g/mol) 203.24
PubChem CID 51885231
Formula Weight 203.24
Percent Purity 98%,98% ee
Physical Form Powder
Chemical Name or Material (S)-α-Methylaspartic acid-4-tert-butyl ester, 98% ee
Show More Show Less
Product Identifier
  • (S)-alpha-Methylaspartic acid-4-tert-butyl ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.